Find methods for your needs
Determination of arsenic in food and beverages with pure oxygen reactive gas using the Thermo Scientific iCAP RQ ICP-MS Be the first to rate this application
Description
To demonstrate the interference free analysis of arsenic in rice and fruit juice after conversion of As into ⁷⁵As¹⁶O+ with pure oxygen reactive gas using the Thermo Scientific iCAP RQ ICP-MS.
Market: Food and Beverage
Keywords: arsenic, Fruit juices, interference removal, oxygen, reaction gases, rice
Matrix: 20 mL microwave digested sample, rice
Author: Bhagyesh Surekar, Chandrakant Pawar, Rakesh Jha, Biswajayee Airaraj Patra
Affiliation: Thermo Fisher Scientific, Mumbai, India
Uploaded on 2/21/2018.
For Research Use Only. Not for use in diagnostic procedures.