Find methods for your needs
U. S. EPA Method 200.7 - Wastewater Analysis for Trace Metals Using an Auto-Dilution System Coupled to the Thermo Scientific iCAP 7000 Plus Series ICP-OES
Description
To demonstrate the ability of the Thermo Scientific iCAP 7000 Plus Series ICP-OES to analyze high concentrated wastewater samples and the coupling of an auto-dilution system for intelligent dilution of samples that are outside the calibration range.| Market: | Environmental; Water Analysis |
| Keywords: | Wastewater analysis, Auto-Dilution, Calibration Range, Effluent Guidelines, EPA Method 200.7, Wastewater Regulations |
| Matrix: | Wastewater |
| Author: | Sanja Asendorf |
| Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.