Find methods for your needs
Analysis of trace elements in naphtha using the Thermo Scientific iCAP 7000 Plus Series ICP-OES
Description
This application demonstrates how volatile organic solvents can be easily analyzed with the Thermo Scientific iCAP 7000 Plus Series ICP-OES fitted with a cooled spray chamber to reduce volatility of the solvent. Cooling down the spray chamber to as low as -10 °C reduces the solvent load on the plasma and allows straight forward analysis of organic solvents with a more stable plasma and reduced background emission.Market: | Petrochem, Oil and Gas; Environmental; Chemical |
Keywords: | Cooled spray chamber, IsoMist, Naphtha, Volatile organic solvents |
Matrix: | naphtha |
Author: | Sanja Asendorf |
Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.