Find methods for your needs
High matrix tolerance of the Thermo Scientific iCAP 7000 Plus Series ICP-OES with the radial Ceramic D-Torch and the sheath gas adaptor
Description
This application note describes how routine analysis of high matrix samples like 25% NaCl can be performed with the Thermo Scientific iCAP 7400 ICP-OES Radial fitted with the additional gas accessory in combination with the Ceramic D-Torch and the sheath gas adaptor.| Market: | Environmental |
| Keywords: | Ceramic D-Torch, high matrix, NaCl, robustness, salt, sheath gas adaptor |
| Matrix: | 25% NaCl |
| Author: | Nora Bartsch |
| Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.