Find methods for your needs
AN72573: Determination of Halogens in Polymers and Electronics using a Combustion Ion Chromatography System
Description
The widespread use of electronic products has drawn increased attention to their impact on the environment. This application note describes a simple CIC method to determine halogens in polymers for electrotechnical products and electronics. Polymer and electronic samples were homogenized by cryogenic grinding, combusted in the Mitsubishi AQF-2100H system, and then directly injected into a Dionex Integrion HPIC system with a Thermo scientific™ Dionex™ IonPac™ AS17-C column set.| Market: | Environmental |
| Keywords: | CIC, AQF-2100H, RFIC System, Dionex Integrion HPIC, Dionex IonPac AS17-C column, IEC 62321-3-2, RoHS, TN72211, AN 72573, TN000767, AN72573, AS17 |
| Matrix: | electronics, polymer |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |