Find methods for your needs
Dionex IonPac AS12A Column Performance Test Using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. This record provides an eWorkflow for executing the QAR method on an HPIC system. See column manual for detail.| Market: | Pharma; Environmental; Chemical; Electronics & Semiconductors |
| Keywords: | AERS 500, Anion, Drug Substance, Oxyhalides, Water, environmental, inorganic anions, RFIC, IC, carbonate eluent, suppressed conductivity detection, QAR, Quality Assurance Report, HPIC IonPac AS12A |
| Matrix: | Drug, Water |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | ThermoFisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.