Find methods for your needs
TN72478: Instrument Configuration for Native N-linked Oligosaccharide Characterization by HPAE-PAD/MS - Gradient 3
Description
The biopharmaceutical industry requires reliable high throughput analysis of N-linked glycans. Many of the commonly used analytical approaches are based on derivatization, which is known to introduce differential loss of sialic acid and incomplete labeling. Preparation of native glycans is not subject to these concerns. This approach starts with a fast in-solution glycoprotein digestion to obtain oligosaccharides. These native glycans are then analyzed by high-performance anion-exchange chromatography with pulsed amperometric detection coupled to MS. This record is for neutral and IgG glycans.| Market: | BioPharma |
| Keywords: | Carbohydrates, Electrochemical detection, Glycans, glycoprotein, asparagine-linked, PAD, IC-MS, MS/MS, HPAE-PAD, Orbitrap, Glycan, complex oligosaccharides, TN 20, TN 70671, TN20, TN70671, TN 72478 |
| Matrix: | glycoprotein oligosaccharides |
| Author: | Zoltan Szabo, J.R. Thayer, and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.