Find methods for your needs
AN72482: Determination of Urea in Ultrapure Water by IC-MS/MS Be the first to rate this application
determination urea
Description
Water used in the production of electronic devices, such as the components that make up computers and mobile phones, must be extremely pure. In particular, it must be free of ionic compounds. Typically, manufacturers aim to have the concentration of each ion below 1 μg/L. Another water contaminant that manufacturers seek to minimize is urea, which has been shown to cause problems with semiconductor production. This application note describes the development and execution of this method for determining urea in UPW.
Market: Electronics & Semiconductors
Keywords: mass spectrometry, MS/MS, semiconductor, HPW, IonPac CS16-Fast, UPW
Matrix: High-purity water
Author: Soon Fatt Lee, Fiona Teh Hui Boon, Chris Cheah Hun Teong, and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 11/20/2017.
For Research Use Only. Not for use in diagnostic procedures.