Find methods for your needs
AN72501: Rapid Determination of Phosphate and Citrate in Carbonated Soft Drinks using Ion Chromatography
Description
This application note updates previous Application Note 169. It presents a new method for the simultaneous determination of phosphate and citrate in carbonated soft drinks using the Dionex IonPac Fast Anion IIIA column. In addition, the sample injection was reduced from 1.2 to 0.4 μL to increase column life.| Market: | Food and Beverage |
| Keywords: | Conductivity Detection, Carbonated beverage, Citrate, RFIC, Cola, ion chromatography, IC, phosphate, beverage analysis, carbonated soft drinks, beverage test, Dionex IonPac Fast Anion IIIA column, soda pop |
| Matrix: | carbonated soft drink |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.