Find methods for your needs
AB72406: Fast Determination of Low Mass, Inorganic Cations in a Ground Water Sample using IC-MS
Description
Reliable analytical methods for food and beverage samples are required in order to report ingredients for food labeling requirements, and to ensure the absence of food contamination. In this application brief, separations of five inorganic cations in a ground water sample were achieved within six minutes. The analytes were detected serially, by suppressed conductivity and single quadrupole mass spectrometry in full scan (FS) and SIM (select ion monitoring) modes. This application updates AN269.Market: | Environmental |
Keywords: | Diethylamine, dimethylamine, Ethylamine, Magnesium, Methylamine, RFIC, Potassium, IonPac CS12A column, IC, Calcium, IC-MS, Sodium, triethanolamine, Ammonium, Single quadrupole mass spectrometer, Integrion IC, ISQ EC mass spectrometer, Monoethanolamine, TN72611 |
Matrix: | 100-fold diluted ground water |
Author: | Terri Christison, Alex Semyonov, and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |