Find methods for your needs
Rapid, sensitive, and easy UHPLC-MS/MS analysis of fungicides in fruit juices with QuEChERS
Description
Quantitation of relevant fungicides in fruit juices down to below 1 ng/mL level without SPE workup. Simplified extraction procedure with QUECHERS sample treatment only. UHPLC-MS/MS analysis with sub-four minute gradients. To develop a rapid and sensitive assay for the quantitation of fungicides in fruit juices (orange and apple) with a separation gradient time of less than four minutes. The QuEChERS method was used without further sample preparation, such as SPE, evaporation, and resuspension, to give lower costs and faster workup.| Market: | Food and Beverage |
| Keywords: | QuEChERS, Orange juice, apple juice, UHPLC, Food Safety, Fruit juice, Quantitation, TSQ Endura, Fungicides, Small molecule, Vanquish Flex Binary UHPLC, Vanquish GOLD |
| Matrix: | Apple juice, Orange juice |
| Author: | Aaron Lamb, Brian King |
| Affiliation: | Thermo Fisher Scientific,Runcorn UK |
For Research Use Only. Not for use in diagnostic procedures.