Find methods for your needs
TN72439: Practical Guidance for Switching between Anion and Cation Analysis Systems: Anions
Description
Municipalities often are required to determine both anions (nitrate, nitrite, fluoride etc) and cations (magnesium, calcium, ammonium) for compliance monitoring. It is ideal to have a dual system that can accommodate both anion and cation analysis. However, many analysts have a single IC system and need to switch between anion and cation analysis. This technical note discusses the operation and installation of the IC system for a smooth transition from an anion to a cation system and vice versa. Anion results are shown in this record. An ICS-6000 can be used for this application.| Market: | Environmental |
| Keywords: | Eluent Generation, ICS-5000+, environmental, anions, RFIC, IC, Water analysis, cations, Anion to cation, cation to anion, HPIC system, ICS-6000 |
| Matrix: | DI water |
| Author: | Manali Aggrawal, Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |