Find methods for your needs
HPLC Assay of Water-Soluble Vitamins, Fat-Soluble Vitamins, and a Preservative in Dry Syrup Multivitamin Formulation Be the first to rate this application
hplc assay watersoluble vitamins fatsoluble vitamins a preservative dry syrup multivitamin formulation
Description
The Acclaim PA2 column can successfully analyze a sample with a mobile phase from 100% aqueous to 100% organic solvent, thereby allowing water- and fat-soluble vitamins to be separated in a single analysis. The Dionex UltiMate 3000 system and Chromeleon software facilitate this analysis by allowing multiple injections during the same run. This method is judged accurate, based on analysis of multivitamin dry syrup and a spiked placebo product. The Acclaim PA2 column, combined with an UltiMate 3000 system, is an excellent solution for vitamin determinations.
Market: Food and Beverage
Keywords: UltiMate 3000, Water-soluble vitamins, Fat-soluble vitamins, multiple injections
Matrix: dry syrup, Water
Author: HPLC Product Marketing
Affiliation: Thermo Fisher Scientific, Germering, Germany
Uploaded on 10/23/2017.