Find methods for your needs
Multi-residue pesticide screening in cereals using GC-Orbitrap mass spectrometry
Description
In this study, the performance of the Thermo Scientific Exactive GC Orbitrap mass spectrometer was evaluated for the routine analysis of GC-amenable pesticides in cereals (wheat, barley, oat, rye, and rice). The Exactive GC-MS system is routinely operated at a resolving power of 60,000 (measured at m/z 200 as full width at half maximum) for the detection of trace compounds against a complex chemical background as encountered in cereal sample extracts.| Market: | Food and Beverage |
| Keywords: | High resolution, Pesticides, QuEChERS, screening, TraceFinder, Quantitation, cereals, GC Orbitrap Mass Spectrometry, Accurate Mass |
| Matrix: | barley, oat, rice, rye, wheat |
| Author: | Dominic Roberts1, Mette Poulsen2 |
| Affiliation: | 1. Thermo Fisher Scientific, 2. University of Denmark |
For Research Use Only. Not for use in diagnostic procedures.