Find methods for your needs
AB72480: Detection of Common Inorganic Anions using a Compact Ion Chromatography System Coupled with a Single Quadrupole Mass Spectrometer
Description
This application brief describes a method for detecting seven common anions using a Thermo Scientific™ Dionex™ Integrion™ HPIC™ system coupled to a Thermo Scientific™ ISQ™ EC single quadrupole mass spectrometer. The separation was performed on a Thermo Scientific™ Dionex™ IonPac™ AS20 column set. With the use of very specific MS SIM detection, ppb-level analyte identification can be achieved with greater confidence on a wide range of samples, such as bottled drinking water.Market: | Environmental |
Keywords: | IonPac AS20 column, IC-MS, Integrion |
Matrix: | Drinking water |
Author: | Beibei Huang;Joshua T. Maze;Kyle B. Renfrew; Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific |
Status: | Proof of Concept |