Find methods for your needs
Identification of metabolites in human plasma using GC-Orbitrap-MS after online derivatisation
Description
In this study, a high resolution accurate mass metabolomics library was used for the identification of metabolites in human plasma. Analysis was performed using a QExactive GC in combination with online sample derivatization. Compound identification was enabled by matching deconvoluted spectra against a high resolution accurate mass metabolomics library containing over 850 metabolites.| Market: | Toxicology; Forensics |
| Keywords: | High resolution, Human Plasma, Metabolites, Orbitrap Technology, Gas Chromatography, QExactive GC, high resolution metabolites library, on-line sample preparation |
| Matrix: | Human plasma |
| Author: | Dominic Roberts, Lana Brockbals, Andrea E. Steuer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.