Find methods for your needs
The determination of elemental impurities in vitamin B12 supplements using the Thermo Scientific iCAP TQ ICP-MS Be the first to rate this application
Description
To demonstrate the accurate determination of elemental impurities, especially As, in vitamin B12 according to the method outlined in USP chapter <233>, Elemental Impurities - Procedures. To demonstrate that triple quadrupole (TQ) ICP-MS can be easy to use and methods can be set up through a dedicated tool, Reaction Finder, which automatically selects optimized parameters with respect to reaction gas and product ion mass.
Market: Pharma; Food and Beverage
Keywords: arsenic, interference removal, oxygen, reaction gases, triple quadrupole, USP 232, USP 233, USP 2232
Matrix: aqueous, vitamin B12 supplement
Author: Daniel Kutscher, Marcus Manecki, Shona McSheehy Ducos,
Affiliation: Thermo Fisher Scientific Bremen, Germany
Uploaded on 9/8/2017.
For Research Use Only. Not for use in diagnostic procedures.