Find methods for your needs
Demonstration of improved efficiency, sensitivity and resolution for the analysis of triazines using a solid core C8 4 µm HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of triazines. The separation is performed on a Thermo Scientific Accucore XL C8 4 µm HPLC column, with UV detection at 220 nm. Compared to a conventional 5 μm fully porous column using the same experimental conditions, the Accucore XL C8 4 μm HPLC column provides improved efficiency, sensitivity and resolution for the analysis of triazines.| Market: | Environmental |
| Keywords: | Accucore XL C8, Ametryn, Atrazine, HPLC, Prometon, Prometryn, Propazine, Simazine, Simetryn, Triazines, UltiMate 3000 RSLC, UV detection |
| Matrix: | N/A |
| Author: | Eilidh MacRitchie |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.