Find methods for your needs
AN72438: Determination of Organic Acids in Animal Feeds using Two Ion Chromatography Methods (Method B)
Description
Organic acids and their salts such as citric acid, malic acid, formic acid, lactic acid, acetic acid, propionic acid, and fumaric acid are animal feed additives which play an important role in animal. In this application note, the animal feed sample is extracted with water. The extract is filtered or centrifuged and diluted, if necessary. The amounts and types of OAs from the sample are then determined using two IC methods. Method A is based on anion-exchange separation and method B is based on ion-exclusion separation. This is Method B. An ICS-6000 can be used for this application.Market: | Food and Beverage |
Keywords: | Dionex ICS-5000+, Dionex IonPac ICE-AS6 ion-exclusion column, suppressed conductivity detection, ICS-6000 |
Matrix: | Water |
Author: | Jingli Hu and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific |