Find methods for your needs
AN72481: Trace Anions by Single Pass AutoNeutralization and Ion Chromatography (IC): DI Water Injection
Description
Trace anions determinations in caustic solutions require high dilution to minimize column overload by the caustic sample. However, the high dilutions required prevent analysis of trace anionic contaminants. Here a single pass approach to neutralize the base sample is demonstrated using both suppressors run in recycle mode. The run time was reduced from 45 min in the previous version of this application, AN 93 using “Park and AutoNeutralize” to 12 min. This application is demonstrated on a DI water sample to determine baseline contamination. An ICS-6000 can be used for this application.| Market: | Chemical |
| Keywords: | AutoNeutralization, caustic, ICS-5000+ HPIC system, fluoride, anions, RFIC, Anion exchange chromatography, IC, Chloride, phosphate, Sulfate, semiconductor, Bromide, IonPac AS28-4µm column, IonPac AS18-4μm column, sulphate, anions in caustic, AN93, electronic chemicals, ICS-6000, AN 72481, AN72481 |
| Matrix: | caustic solutions, DI water baseline |
| Author: | Soon Fatt Lee, Aaron Rose, Terri Christison, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.