Find methods for your needs
Rapid analysis of fat soluble vitamins using a Thermo Scientific Accucore RP-MS HPLC column Be the first to rate this application
rapid analysis fat soluble vitamins using a thermo scientific accucore rpms hplc column
Description
The Thermo Scientific HPLC system is applied for the analysis of fat soluble vitamins. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 3 minutes, with UV detection at 280 nm.
Market: Food and Beverage
Keywords: Accucore RP-MS, Fat soluble vitamins, HPLC, UV detection, Vitamin A, Vitamin D3, Vitamin E, Vitamin E acetate, Vitamin K
Matrix: N/A
Uploaded on 8/24/2012.
For Research Use Only. Not for use in diagnostic procedures.