Find methods for your needs
AN346: Totally Automated Sample Preparation Using Accelerated Solvent Extraction (ASE) Coupled with Gilson ASPEC: The Determination of Dianthrones in St. John's Wort
Description
This Application Note reports on the use of a coupled ASE-ASPEC system as an automated extraction and extract manipulation device for the determination of dianthrones in St. John’s Wort (Hypericum perforatum). The results using the coupled system are compared to those obtained using a traditional sonication procedure followed by manual SPE cleanup.| Market: | Pharma |
| Keywords: | ASE, ASE 200, ASE 350, Dianthrones, St. John's Wort |
| Matrix: | St. John's wort |
For Research Use Only. Not for use in diagnostic procedures.