Find methods for your needs
AB876: Automated Solid-Phase Extraction (SPE) of Wastewaters and Surface Watersfor Polyaromatic Hydrocarbons and Phthalates — Modification of EPA Method 625
Description
EPA Method 525.2, Determination of Organic Compounds in Drinking Water by Liquid-Solid Extraction and Capillary Column Gas Chromatography/Mass Spectrometry, can be used as a basis for developing a liquid-solid extraction method for EPA Method 625, Methods for Organic Chemical Analysis of Municipal and Industrial Wastewater. Unlike the drinking water methods, however, analysis of wastewaters need to contend with particulates. The AutoTrace™ 280 Solid-Phase Extraction (SPE) instrument has features that allow analysis of samples even with high amounts of suspended solids.| Market: | Environmental |
| Keywords: | PAHs, Solid phase extraction, SPE, Polyaromatic hydrocarbons, AutoTrace 280 |
| Matrix: | Surface water, Waste water |
| Author: | David Larabee-Zierath |
| Affiliation: | University Hygienic Laboratory, Iowa’s Environmental and Public Health Laboratory, Iowa City, IA |
For Research Use Only. Not for use in diagnostic procedures.