Find methods for your needs
AB805: Automated Solid Phase Extraction (SPE) of Dioxins and Furans in Surface Water
Description
The EPA Methods 8260 and 8270 are the basis of the sample preparation of polychlorinated dibenzo-p-dioxins, polychlorinated dibenzofurans and PCBs in surface and ground water. The SPE method described in this Application Brief (AB) also recovers these compounds along with 1,4-Dioxan.| Market: | Environmental |
| Keywords: | Solid phase extraction, SPE, EPA 8270, PCBs, AutoTrace 280, automated SPE, dibenzofurans, dibenzo-p-dioxins, EPA 8260 |
| Matrix: | Surface water |
For Research Use Only. Not for use in diagnostic procedures.