Find methods for your needs
AB806: Automated Solid Phase Extraction (SPE) of Nitrosamines in Water
Description
Due to the carcinogenic nature of nitrosamines at very low levels, new regulations setting detection limits at PPT (ng/L) levels have been set for nitrosamines in drinking water, milk, beer, and food products. EPA Method 521 recommends coconut carbon SPE cartridges and this method can be easily adapted for the analysis of food and beverages.| Market: | Environmental |
| Keywords: | SPE, Diphenylamine, NDMA, NMEA, NDEA, NDPA, NMOR, NPYR, NPIP, NDBA, DPhA, methylethylnitrosamine, nitrosodiethylamine, nitrosodi-n-propylamine, nitrosopyrrolidine, nitrosopiperidine, nitrosodi-n-butylamine, nitrosomorpholine, Nitrosodimethylamine, AutoTrace 280, carbon-based SPE |
| Matrix: | Drinking water |
For Research Use Only. Not for use in diagnostic procedures.