Find methods for your needs
AN359: Extraction of Contaminants, Pollutants, and Poisons from Animal Tissue Using Accelerated Solvent Extraction (ASE)
Description
Accelerated Solvent Extraction (ASE) uses solvents at elevated temperatures and pressures to extract organic materials from solid and semisolid samples in a fraction of the time required by traditional extraction procedures. This Application Note details procedures for extracting the following contaminants from animal tissues: Dioxins/Furans, Polybrominated Flame Retardants (PBDE), PCBs, pesticides, PAHs, and organo-tin.| Market: | Food and Beverage |
| Keywords: | PAHs, PBDE, PCB, ASE, ASE 200, ASE 350, PCBs, oyster, PCB Congener, Animal tissue |
| Matrix: | Animal tissue |
For Research Use Only. Not for use in diagnostic procedures.