Find methods for your needs
ASTM D6352 simulated distillation of high boiling point hydrocarbons using Thermo Scientific TraceGOLD TG-1MT Metal GC Column
Description
The Thermo Scientific Trace GC system is applied for the ASTM D6352 simulated distillation of high boiling point hydrocarbons. The separation is performed on a Thermo Scientific TraceGOLD TG-1MT 6m x 0.53mm (0.15µm film), with flame ionisation detection.| Market: | Chemical |
| Keywords: | ASTM D6352, Hydrocarbons, simulated distillation, TG-1MT |
| Matrix: | Standard |
For Research Use Only. Not for use in diagnostic procedures.