Find methods for your needs
AN343: Determination of Pesticides in Large-Volume Food Samples Using Accelerated Solvent Extraction (ASE)
Description
Accelerated Solvent Extraction (ASE) is an extraction technique that speeds the extraction process and reduces the total amount of solvent used. The system uses conventional solvents at elevated temperatures and pressures, which results in improved extraction kinetics. The Dionex ASE 300 has the capability to extract samples with volumes as large as 100 mL. This application note reports on the use of the ASE 300 for the determination of organophosphorus pesticides (OPPs) in fruits and vegetables.| Market: | Food and Beverage |
| Keywords: | Parathion-ethyl, Pesticides, ASE, Diazinon, Malathion, Chlorpyrifos, Mevinphos, Phorate, Dimethoate, Disulfoton, Fenchlorphos, Fenthion, EPN, Azinphos-methyl, Coumaphos, ASE 350, ASE 300, Food Samples, Dichlorvos/Naled, TEPP, Demeton-O, Ethoprophos (Ethoprop), Sulfotep, Demeton-S, Parathion-methyl, Trichloronat, Tetrachlorvinphos, Prothiofos, Merphos, Fensulfothion, Sulprofos |
| Matrix: | Foods |
For Research Use Only. Not for use in diagnostic procedures.