Find methods for your needs
AN324: Accelerated Solvent Extraction of Hydrocarbon Contaminants (BTEX, Diesel, and TPH) in Soils
Description
Accelerated solvent extraction is an innovative sample preparation technique that combines elevated temperatures and pressures with liquid solvents to achieve fast and efficient removal of analytes of interest from various matrices. Accelerated solvent extraction has been demonstrated to be equivalent to existing extraction methodologies such as Soxhlet and automated Soxhlet for most RCRA (Resource Conservation and Recovery Act) analytes from solid and semisolid samples. It meets the requirements of U.S. EPA Method 3545, Pressurized Fluid Extraction.| Market: | Environmental |
| Keywords: | ASE, Soils, ASE 200, ASE 350, EPA 3545, Hydrocarbon Contaminants |
| Matrix: | Soil |
| Author: | Bruce Richter |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.