Find methods for your needs
AN341: Extraction of Base/Neutrals and Acids (BNAs) from Large-Volume Samples Using Accelerated Solvent Extraction (ASE)
Description
The procedures described in this application note meet the requirements for sample extraction as determined by U. S. EPA Method 3545(A). This method is applicable to the extraction of water-insoluble or slightly water-soluble volatile and semivolatile compounds in preparation for GC/MS measurement. This method is applicable to soils, clays, wastes, and sediments containing from 250–12500 µg/kg of BNA compounds.| Market: | Environmental |
| Keywords: | 2,4,6-Trichlorophenol, 2,4-Dichlorophenol, 2,6-Dinitrotoluene, 2-Chlorophenol, 4-Nitrophenol, Acenaphthene, Anthracene, Benzidine, Benzyl alcohol, Chrysene, Fluorene, Naphthalene, Nitrobenzene, Phenanthrene, Phenol, Pyrene, Pyridine, ASE, Hexachlorobutadiene, Hexachlorobenzene, ASE 350, ASE Prep DE, Soxhlet Extraction, 2-Fluorobiphenyl, sonication, U. S. EPA Method 3545(A), N-Nitrosodimethylamine, Aniline, Bis(2-chloroethyl)ether, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, 1,2-Dichlorobenzene, 2-Methylphenol, Bis(2-chloroisopropyl)ether, 4-Methylphenol, N-Nitroso-di-n-propylamine, Hexachloroethane, Isophorone, 2,4-Dimethylphenol, 2-Nitrophenol, Bis(chlorethoxy)methane, 1,2,4-Trichlorobenzene, 4-Chloroaniline, 4-Chloro-3-methylphenol, 2-Methylnaphthalene, Hexachlorocyclopentadiene, 2,4,5-Trichlorophenol, 2-Chloronaphthalene, 2-Nitroaniline, Azobenzene, Bis(2-ethylhexyl)phthalate, Dimethylphthalate, Diethylphthalate, Acenaphthalene, 3-Nitroaniline, 4,6-Dinitro-2-methylphenol, 2,4-Dinitrotoluene, Dibenzofuran, 4-Chlorophenyl-phenylether, 4-Nitroaniline, N-Nitrosdiphenylamine, 4-Bromophenyl-phenylether, 7 Pentachlorophenol, Carbazole, 40 Fluoranthene, 3,3'-Dichlorobenzidine, Benz[a]anthracene, Benzo[b]fluoranthene, Benzo[k]fluoranthene, Benzo[a]pyrene, Indeno[1,2,3-cd]pyrene, Dibenz[a,h]anthracene, Benzo[g,h,I]perylene, 8 Di-n-butylphthalate, Butylbenzylphthalate, Di-n-octylphthalate |
| Matrix: | Clays, Sediments, Soils, Wastes |
For Research Use Only. Not for use in diagnostic procedures.