Find methods for your needs
AN355: Rapid Extraction and Determination of Arsenicals in Fish Tissue and Plant Material Using Accelerated Solvent Extraction (ASE)
Description
The toxicity of arsenic is species dependent. Inorganic arsenic species such as arsenite (As[III]) and arsenate (As[V]) have been classified as carcinogens. Methylated forms such as monomethylarsonic acid (MMA) and dimethylarsinic acid (DMA) have recently been labeled as cancer promoters. Arsenobetaine (AsB), arsenocholine (AsC), and arseno sugars have been found to be relatively nontoxic. This application note describes ASE methods for the extraction of various arsenic species from different food matrices, specifically fish tissue, ribbon kelp, and vegetables grown in contaminated soil.| Market: | Food and Beverage |
| Keywords: | Arsenate, Arsenite, MMA, Accelerated solvent extraction, ASE, Arsenobetaine, AsB, As(III), As(V), DMA, IC-ICP-MS, ASE 200, ASE 350, IC-ICP/MS, arsenic species, As m/z 328, As m/z 428, As m/z 392, arseno sugar, arsenosugar, Monomethylarsonic acid, monomethyl arsenate, dimethylarsenic acid, dimethyl arsenate, kelp, fish tissue, carrots, arsenic speciation, total arsenic |
| Matrix: | Carrots, Freeze dried fish tissue, Kelp |
For Research Use Only. Not for use in diagnostic procedures.