Find methods for your needs
ASTM D7213 analysis using Thermo Scientific TraceGOLD TG-1MT Metal GC Column
Description
The Thermo Scientific Trace GC system is applied for the ASTM D7213 simulated distillation of petroleum fractions in the 100 to 615°C boiling point range. The separation is performed on a Thermo Scientific TraceGOLD TG-1MT 6m x 0.53mm (0.15µm df), with flame ionisation detection.| Market: | Chemical |
| Keywords: | ASTM D7213, Fractions, Hydrocarbons, petroleum, simulated distillation, TG-1MT |
| Matrix: | Standard |
For Research Use Only. Not for use in diagnostic procedures.