Find methods for your needs
AN330: Accelerated Solvent Extraction (ASE) of the Organic Components of Granular and Liquid Detergents
Description
Organic solvent extraction is a standard technique in the detergent industry for isolation of organic components from inorganic matrices and separation of ionic and nonionic surfactants. For example, analysis of the alcohol-soluble portion of detergents is routinely performed to verify the integrity of the manufacturing process. Traditional extraction of this material is labor-intensive and uses large amounts of liquid solvent. With ASE, manufacturers can streamline the extraction process to quickly verify product quality and provide feedback information into the production cycle.| Market: | Chemical |
| Keywords: | Detergent, ASE, ASE 200, ASE 350, IR, gravimetric analysis, Liquid detergent, granular detergent |
| Matrix: | Detergents |
For Research Use Only. Not for use in diagnostic procedures.