Find methods for your needs
AN316: Extraction of PCBs from Environmental Samples Using Accelerated Solvent Extraction
Description
Accelerated solvent extraction is an extraction method that significantly streamlines sample preparation. PCBs are found in many solid waste materials. This AN describes the application of accelerated solvent extraction to the extraction of PCBs from sewage sludge, river sediments, marine sediments, and oyster tissue. The procedures described in this AN meet the requirements for sample extraction of solid samples as determined by U.S. EPA Method 3545.| Market: | Environmental |
| Keywords: | GC, GC-MS, PCB, ASE, ASE 200, ASE 350, U.S. EPA Method 3545 (SW-846 Update III), PCBs, Sewage Sludge, marine tissue, marine sediment, oyster, river sediment, PCB Congener |
| Matrix: | Sewage sludge |
| Author: | John Ezzell and Bruce Richter |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.