Find methods for your needs
AN326: Extraction of Drugs from Animal Feeds Using Accelerated Solvent Extraction (ASE)
Description
ASE is widely used in the environmental industry to replace time- and solvent-intensive techniques such as Soxhlet and sonication extraction. Many features of the ASE system also make it attractive for use in pharmaceutical laboratories. This Application Note gives two examples of how ASE can provide extraction efficiencies superior to other techniques. In the first example, ASE is used to extract an antischizophrenic agent from rodent feed used in drug testing. In the second example, ASE is used to extract Lasalocid, a veterinary medicinal added to poultry and cattle feed.| Market: | Pharma |
| Keywords: | Accelerated solvent extraction, ASE 350, veterinary medicinal, Animal Feeds, cattle feed, Antischizophrenic Agent, Poultry and Cattle Feed, Lasalocid |
| Matrix: | Animal feeds |
For Research Use Only. Not for use in diagnostic procedures.