Find methods for your needs
AN320: Extraction of Chlorinated Pesticides Using Accelerated Solvent Extraction (ASE)
Description
Accelerated Solvent Extraction (ASE®) is an extraction method that significantly streamlines sample preparation. The procedures described in this application note meet the requirements for sample extraction as prescribed by EPA Method 3545. This method is applicable to the extraction of water-insoluble or slightly water-soluble volatiles and semivolatiles in preparation for gas chromatographic or GC/MS measurement. The method is applicable to the extraction of chlorinated pesticides from soils, clays, wastes, and sediments containing from 5 to 250 μg/kg of the target compounds.| Market: | Environmental |
| Keywords: | Chlorinated pesticides, Accelerated solvent extraction, Soils, ASE 350, sludges, EPA Method 3545 |
| Matrix: | Soil samples |
For Research Use Only. Not for use in diagnostic procedures.