Find methods for your needs
AN313: Extraction of PAHs from Environmental Samples by Accelerated Solvent Extraction (ASE)
Description
Accelerated Solvent Extraction (ASE) is a new extraction method that significantly streamlines sample preparation. ASE provides a more convenient, faster, and less solvent intensive method than previously available for extraction of PAHs from solid wastes. PAH recoveries by ASE are equivalent to other more solvent intense methods such as Soxhlet or sonication. ASE also avoids the multiple washing procedures associated with sonication. The procedures described here meet the requirements for extraction of PAHs from solid waste in U.S. EPA Method 3545.| Market: | Environmental |
| Keywords: | ASE, Soils, Polyaromatic hydrocarbons, ASE 200, ASE 350, EPA 3545, sediments, sludges, solid waste |
| Matrix: | Soil |
For Research Use Only. Not for use in diagnostic procedures.