Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the assay analysis of sodium lactate. The separation was performed on a Thermo Scientific Acclaim 120-C18 HPLC column with a UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, UV detection, Sodium Lactate |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.