Find methods for your needs
Separation of related impurities from Icatibant (Peptidomimetic compound)
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied to the separation of the related impurities of Icatibant. The separation was performed on a Thermo Scientific Hypersil Gold HPLC column with UV detection at 210 nm. With a total run time of 75 minutes the method allows separation of all related impurities of Icatibant.| Market: | Pharma |
| Keywords: | HPLC, Hypersil GOLD, UV detection, Impurity profile, Icatibant |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.