Find methods for your needs
AN1086: Automated Solid Phase Extraction of 14 Explosives in Tap Water Based on U.S. EPA Method 8330 Using Hydrophilic Reversed-Phase Cartridge Be the first to rate this application
spe explosives tap water
# Compound Name Compound Class RT (min)
1 RDX Explosive 14.00
2 1,3,5-trinitrobenzene Explosive 15.00
3 1,3-Dinitrobenzene Explosive 18.00
4 Nitrobenenze Explosive 19.00
5 2,4,6-trinitrobenzene Explosive 24.00
6 Tetryl Explosive 25.00
7 2,6-Dinitrotoluene Explosive 27.00
8 2,4-Dinitrotoluene Explosive 28.00
9 2-Nitrotoluene Explosive 30.50
10 4-Nitrotoluene Explosive 34.00
11 3-Nitrotoluene Explosive 36.50
12 4-Amino-2,6-Dinitrotoluene Explosive 39.00
13 2-Amino-4,6-Dinitrotoluene Explosive 42.00
Description
The U.S. EPA Method 8330, first introduced in November 1990, is the most common method for the analysis of explosives. Analytes can be detected down to 2.5 ppb in water, soil, or sediment. Used here are the Thermo Scientific Dionex AutoTrace 280 Solid-Phase Extraction (SPE) instrument and the Thermo Scientific Dionex SolEx SPE HRPHS cartridge to concentrate the explosive residues, followed by HPLC.
Market: Environmental
Keywords: 1,3,5-TNB, 1,3-DNB, 2,4,6-TNT, 2,4,6-Trinitrotoluene, 2,4-DNT, 2,6-Dinitrotoluene, 2,6-DNT, 2-NT, 3-NT, 4-Nitrotoluene, 4-NT, EPA Method 8330, Nitrobenzene, Tetryl, HMX, RDX, NB, AutoTrace 280 SPE, Drinking water, SolEx HRPHS, 1,3,5-trinitrobenzene, 4-amino-2,6-dinitrotoluene, 2-amino-4,6-dinitrotoluene, 2,4-Dinitrotoluene, Nitro compounds, Automated off-line SPE, 2-Am-DNT, 4-Am-DNT, 2-Nitrotoluene, 3-Nitrotoluene, 1,3-Dinitrobenzene, Octahydro-1,3,5,7-tetranitro- 1,3,5,7-tetrazocine, Methyl-2,4,6- trinitrophenylnitramine, Hexahydro-1,3,5- trinitro-1,3,5-triazine
Matrix: Drinking water
Author: Jing Hong and Rosanne Slingsby
Affiliation: Thermo Fisher Scientific
Uploaded on 7/15/2016.
For Research Use Only. Not for use in diagnostic procedures.