Find methods for your needs
AN1080: Automated Solid-Phase Extraction of Linear Alkylbenzene Sulfonatein Wastewater Using a Weak Anion-Exchange Cartridge Followed by HPLC with UV Detection
Description
Surfactants are the major active ingredients of laundry detergents. Anionic surfactants account for 60% of surfactant used in the U. S. Linear alkylbenzene sulfonate (LAS) is an anionic surfactant. It is a mixture of closely-related isomers and homologues, each containing an aromatic ring sulfonated at the para position and attached to a linear alkyl chain. This application provides a complete workflow method for the extraction and analysis of LAS is waste water samples.| Market: | Environmental |
| Keywords: | Solid phase extraction, SPE, Surfactant, AutoTrace 280, SolEx SPE WAX Cartridges |
| Matrix: | Waste water |
| Author: | Jing Hong and Rosanne Slingsby |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.