Find methods for your needs
AN1097: Determination of Pesticide Residues in Drinking Water Using Automated Solid-Phase Extraction and Gas Chromatography with Nitrogen Phosphorus Detection
Description
This application note addresses the detection of trace amounts of organophosphorus pesticides in water and optimizes the conditions for solid-phase extraction (SPE) and chromatographic analysis using the AutoTrace 280 Solid-Phase Extraction instrument and the Thermo Scientific™ TRACE™ 1300 Series gas chromatograph with a nitrogen-phosphorus detector (NPD).| Market: | Environmental |
| Keywords: | Solid phase extraction, SPE, Methyl parathion, Malathion, Dichlorvos, Phorate, Dimethoate, Disulfoton, Parathion, Drinking water, TRACE 1310 GC, AutoTrace 280, Chinese Method GB 5750.9-2006, GC-NPD: Organophosphorous Pesticides, Demeton |
| Matrix: | Drinking water (spiked) |
| Author: | Liu Qian, Zheng Hongguo, and Yu Chongtian |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.