Find methods for your needs
CAN121: Determination of Polychlorinated Biphenyls (PCBs) in Soils and Solid Waste by Accelerated Solvent Extraction and GC-MS/MS
Description
Polychlorinated biphenyls (PCBs) belong to a broad family of synthetic organic chemicals known as chlorinated hydrocarbons. In the United States, PCBs were produced from 1929 until their manufacture was banned in 1979.Although all 209 of the PCB congeners can be synthesized, only about 130 individual congeners have been identified in commercial PCB mixtures at concentrations ≥ 0.05%. The method reported here is applicable for the determination of 32 of the 209 possible PCB congeners in soils, including the dioxin-like PCB (“non-ortho”,“mono-ortho PCB”, and “di-ortho PCB”).| Market: | Environmental |
| Keywords: | GC, PCB, TraceGOLD TG-17MS, ASE, Rocket Evaporator, ASE 200, PCB Congener, ASE 350, GC/MS/MS |
| Matrix: | Soils, Solid waste |
| Author: | Luca Teli and Fabrizio Galbiati* |
| Affiliation: | Consulenze Ambientali SpA and *Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.