Find methods for your needs
AN1109: Rapid Determination of Nitrobenzenes in Drinking Water Using Automated SPE with GC-ECD
Description
Nitrobenzenes are precursors for the production of numerous industrial chemicals and may be released into the environment through wastewater discharge and may contaminate ground and surface waters. This AN demonstrates automated SPE with GC-ECD for the determination of 15 nitrobenzene compounds in drinking water. The automated Thermo Scientific™ Dionex™ AutoTrace 280 SPE instrument was used in conjunction with the Thermo Scientific™ TRACE™ 1310 GC.This work optimizes SPE and GC conditions and the results show high sensitivity for the 15 nitrobenzene compounds specified in HJ 648-2013.| Market: | Environmental |
| Keywords: | GC, AutoTrace 280 SPE, Drinking water, GC-ECD, Method HJ 648-2013, TRACE 1310 GC |
| Matrix: | Drinking water |
| Author: | Liu Qian |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.