Find methods for your needs
High Precision Pesticide Analysis in Produce using GC Triple Quadrupole and U-SRM Mode
Description
Pesticides are widely used in agriculture to protect crops and to improve efficiency of production. Consequently, governments, food producers and food retailers have the duty to ensure that any residues occurring in foods for human consumption are at or below Statutory Maximum Residue Levels (MRLs). Regulation EC 396/2005 adopted in the European Union sets MRLs for more than 500 different pesticides in over 300 different food commodities. We describe a method providing high quality and low level analysis of pesticides in produce samples using the TSQ Quantum XLS Ultra GC-MS/MS system.| Market: | Food and Beverage |
| Keywords: | Selectivity, Pesticide Analysis, 1 ppb Levels, Challenging Compounds, PTV Backflush, U-SRM Mode, Selected Reaction Monitoring, Timed-SRM |
| Matrix: | Produce |
| Author: | Inge de Dobbeleer, Joachim Gummersbach, Hans-Joachim Huebschmann, Anton Mayer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.