Find methods for your needs
Characterization of Triacylglycerides (TAGs) in Vegetable Oils using MALDI LTQ Orbitrap XL Instrumentation
Description
The world’s population growth has led to increase in consumption of vegetable oils by 4.5% in 2011. By 2020, the annual production is expected to expand by 23%. Import rates for vegetable oils into the EU are predicted to rise significantly. Cases of economic adulteration of vegetable oils with non-authentic and less expensive oil adjuncts occur frequently. We describe a method to uncover structures of specific TAGs and TAG distribution patterns among various vegetable oils exploiting full MS and CID-based MSn experiments using the Thermo Scientific MALDI LTQ Orbitrap XL.Market: | Food and Beverage |
Keywords: | High resolution, Food Safety, Accurate mass, Structural elucidation, MALDI LTQ Orbitrap XL, Orbitrap Technology, MSn, triacylglycerides (TAGs), saturated and unsaturated TAGs, Matrix-Assisted Laser Desorption/Ionization, structural confirmation |
Matrix: | Vegetable oils |
Author: | Chris Weise, Tabiwang Arrey, Kerstin Strupat |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.