Find methods for your needs
Quantitative Analysis of Environmental Air Contaminants Using APCI-MS/MS in Mobile Laboratories
Description
There are many potential hazards in our environment. Chemical emissions, accidental chemical spills and fires are of particular concern. A real-time analytical atmospheric pressure chemical ionization-tandem mass spectrometry (APCI-MS/MS) method for the quantitative analysis of air contaminants has been developed using a customized, direct-sampling APCI device coupled with a Thermo Scientific TSQ series triple stage quadrupole mass spectrometer.| Market: | Environmental |
| Keywords: | Environmental Monitoring, TSQ Series Triple Quadrupole MS, APCI-MS/MS |
| Matrix: | Air |
| Author: | Germain Tremblay, Lise Blanchard, Dominic Lortie, Calin G. Znamirovschi |
| Affiliation: | Ministère du Développement durable, de l’Environnement et des Parcs-Canada; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.