Find methods for your needs
Screening Method for 30 Pesticides in Green Tea Extract Using Automated Online Sample Preparation with LC-MS/MS
Description
Analysis of pesticide residues has been one of the most important tasks of food safety laboratories. Mass spectrometers (MS), with liquid chromatography coupled to triple stage quadrupole mass spectrometers (LC-MSMS), have been the main tools used in pesticide residue analysis. There is a consensus that sample preparation is becoming the bottleneck to the entire workflow. We developed a rapid and sensitive automated online sample preparation LC-MS/MS method to screen for multiple pesticides in green tea extract.| Market: | Food and Beverage |
| Keywords: | Pesticides, Food Safety, TurboFlow Technology, Aria OS Software |
| Matrix: | Green tea |
| Author: | Yang Shi, Catherine Lafontaine, Fangting Ye, Zheng Jiang, Le Ma, Ting Liu, Haijian Wang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.