Find methods for your needs
Fast Screening and Quantification of Pesticide Residues Using a Comprehensive LC-MS Solution
Description
We present a multi-residue analysis method enabled by Thermo Scientific Pesticide Explorer Collection Standard Quantitation Solution. LC–triple-stage MS provides rapid and robust quantitation of more than 250 pesticides below their required MRL. This comprehensive solution includes the QuEChERS sample preparation kit, Dionex UltiMate 3000 LC system, TSQ Endura triple quadrupole MS, TraceFinder software, Accucore aQ column. Method parameters provide a start-to-finish workflow for pesticide analysis. Results address stringent EU guidelines concerning sensitivity, accuracy and precision.| Market: | Other |
| Keywords: | QuEChERS, Pesticide Explorer Collection, European Regulation 396/2005, Commission Directive 2006/125/EC, European Commission 2002/657/EC, SANCO/12571/2013, European Commission 788/2012/EC |
| Matrix: | Foods |
| Author: | Katerina Bousova, Ebru Sarikaya, Michal Godula, Claudia Martins, and Ed George |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.