Description
本文采用水系滤膜和On-Guard RP 柱对洗虾粉未知样品进行净化,首先利用离子色谱分离及质谱的强大定性能力,将二者联用确定了样品中含有硫代硫酸盐和焦磷酸盐,再用离子色谱法测定了样品中的硫代硫酸盐和焦磷酸盐,其色谱峰分离效果好,方法稳定性强、灵敏度高、干扰少,满足对于这种洗虾粉粉末未知成份的检测与确证要求。将本方法用于对食品加工中非法添加物的确证及检测具有十分重要的意义。| Market: | Food and Beverage |
| Keywords: | 离子色谱;质谱;洗虾粉;未知成份 |
| Matrix: | 洗虾粉 |
| Author: | 胡忠阳 |
For Research Use Only. Not for use in diagnostic procedures.